Learn More
Cytiva SAMPLE LOOP 1.0 ML, INV-907
Supplier:  Cytiva 18111401
Encompass
Sample loop for use with �KTA systems. Wetted materials are PEEK. Tubing ID 0.75 mm, OD 1/16" fitted with fingertight connectors 1/16".0
Catalog No. 50-105-5654