Learn More
MilliporeSigma™ Supelco™ Sample Loop for Model 7725 Injector, Stainless Steel
Sample loop for model 7725 injector (volume 100 μL) is best suited for advanced fluid-handling solutions for broad range of analytical instrumentation and in vitro diagnostic systems. It is useful in high performance liquid chromatographic (HPLC) systems and is quite accurate and precise in injecting 1 μL-5 mL concentration of samples.
Supplier: MilliporeSigma™ Supelco™ 57632
Description
Specifications
Stainless Steel | |
Model 7725 | |
5 mL |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.