Learn More
Cytiva (Formerly Pall Lab) Jumbosep™ Spare Parts and Accessories - Sample reservoir and cap (12/pkg)
Supplier:  Cytiva FD002X65
Encompass
Convenient and Reliable Concentration, Purification, and Diafiltration of 20 to 60 mL of Biological Samples-Typically concentrates 60 mL sample volumes to 5 mL in 30 minutes-Achieve 50X concentration and > 90% recovery in just minutes-Low protein-binding Omega™ membrane and polysulfone housing minimize losses due to non-specific binding-Versatile Omega membrane is available in a variety of MWCOs-Color-coded and laser etched for easy identification -Built-in deadstop prevents spinning to dryness-Unique sealing mechanism prevents retentate leakage and filtrate contamination-Economical - Sample reservoir and filtrate receiver can be sanitized or autoclaved, and reused- Omega (modified polyethersulfone) membrane Insert sold separately This product is not approved for use in medical, clinical, surgical or other patient applications.
Catalog No. 50-206-3445