Learn More
SPEX™ SamplePrep DNAmax Isolation Kit
Description
- The complete kit solution for DNA extraction of animal and plant tissue
- Reinforced tubes designed for homogenization through rapid beat beating
- Silica-based columns capture DNA and allow for its purification
- Protocol and workflow optimized for Genomax, Geno/Grinder, and GenoLyte
Specifications
Specifications
| Buffer | Lysis, Wash, Elution |
| Bead Type | Stainless Steel |
| Sample Type | Soft Plant Tissue |
| For Use With (Equipment) | Genomax, Geno/Grinder, GenoLyte |
| For Use With (Application) | DNA Extraction of Animal and Plant Tissue |
| Includes | Lysis Buffer, Binding Buffer B1, Wash Buffer PW1, Wash Buffer PW2, Elution Buffer, Proteinase K Powder, Rnase A, and Pre-Loaded Tubes with Five 3 mm 440 C Stainless Steel Balls |
| No. of Reactions | 75 |
| Product Type | DNA Isolation Kit |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.