Learn More
Qaqc Lab LLC SAMPLER THIEF DISP 100ML 20/CS
Supplier:  Qaqc Lab LLC 6758073H500
Encompass_Preferred
DISPOSABLE SLOT SAMPLERS HDPE; Non-sterile; 3 slots; volume: 100mL; length: 620mm; diameter: 5 mm; 20 per case
Catalog No. NC1559484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.