Learn More
Research Products International Corp Sampling Scp S XL 500ml 10pk
SDP
Supplier:  Research Products International Corp 249510
Encompass_Preferred
"Durable Sampling Scoops are designed for easy sample retrieval of granules, powders and samples.Disposable,sterile scoops are idea for single use and prevent cross-contamination. Scoops feature large, wide openings for easy sample retrieval; flat bottom to stabilize sample on scale;shatterproof, BSE/TSE free. Maufactured from FDA food grade polystyrene in an ISO8 Class clean room; meet quality control and GMP requirements for sampling.Durable designSterile, Individually wrappedSafe sample recovery Electron beam sterilized and individually wrapped.Certificate of Sterility available upon request.Dimensions: 10.0 x 3.6 x 2.1 inches Research or further manufacturing use only, not for food or drug use."
Catalog No. 50-238-8373
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.