Description
- Four PVC legs connected with two PVC T-connectors
- Lightweight, plastic device cleanly and quickly marks off either one-quarter square meter (0.25m2) or one square meter (1.0m2)
- When unfolded, it pops into place using a durable shock cord inside the PVC pipe legs
- T-connectors allow the frame to sink for use in water studies
- Collapsible for transport and storage
- Shock cord is contained inside the pipe legs
Specifications
Specifications
| Size | 1 m |
| For Use With (Application) | Ideal to quantify the results of high school field studies and can be used on land or in water |
| Includes | 4 PVC legs, 2 PVC T connectors, shock cord |
| Product Type | Sampling Square |