Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL BioMedical SampLok™ Adapter Cap 3

Catalog No. 22799902 Shop All ITL BioMedical Products
Change view
Click to view available options
Quantity:
600/Cs., 100/Box, 6 Boxes/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-799-902 600/Cs., 100/Box, 6 Boxes/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-799-902 Supplier ITL BioMedical Supplier No. A100760
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Small and mighty. For blood culture sample collection; compact, simple and efficient.

  • Compact size
  • Optimal for one handed process
  • Secure fit with a range of bottles
  • Reduced biohazard waste and related costs
  • Optional insert available for use with blood collection tubes
  • Compliance

    CE marked

    Specifications

    Product Type Adapter Cap
    Pre-sterilized No
    Autoclavable Autoclavable
    Description Not Made with Natural Rubber Latex
    Material Polypropylene
    Color Clear
    For Use With (Application) Blood Collection
    Quantity 600/Cs., 100/Box, 6 Boxes/Cs.
    Sterility Non-sterile
    For Use With (Equipment) Blood Collection Needles, Blood Culture Bottle, Blood Collection Tubes
    Disposable Single-use
    Show More Show Less
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.