Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ITL BioMedical SampLok™ Adapter Cap 3 Insert

Catalog No. 22799903 Shop All ITL BioMedical Products
Change view
Click to view available options
Quantity:
900/Cs., 150/Box, 6 Boxes/Cs.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-799-903 900/Cs., 150/Box, 6 Boxes/Cs.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 22-799-903 Supplier ITL BioMedical Supplier No. A100761
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A companion product with SampLok™ Adapter Cap 3 when collecting blood sample tubes after collecting blood culture bottles for a more secure fit

  • Optional companion insert for use with SampLok™ Adapter Cap 3
  • Optional insert for use with sample tubes and long-neck culture bottles, can be attached to blood collection and winged needle sets
  • Compliance

    CE marked

    Specifications

    Pre-sterilized No
    Product Type Adapter Cap Insert
    Autoclavable Autoclavable
    Material Polypropylene
    Color Clear
    For Use With (Application) Blood Collection
    Quantity 900/Cs., 150/Box, 6 Boxes/Cs.
    Sterility Non-sterile
    For Use With (Equipment) SampLok™ Adapter Cap 3, Blood Collection Tubes, Blood Collection Needles, Long-neck Culture Bottles
    Product Title
    Select an issue

    By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.