Learn More
Description
- Reduces the number of procedure steps, streamlining and simplifying the collection and transfer of biological samples
- SampLok™ Sampling Kit can be used with various culture bottles and vacuum tubes
- Protect staff from needlestick injuries
Specifications
Specifications
| Product Type | Collection and Transport System |
| Format | Tube |
| Sterility | Sterile |
| Closure Type | Inegrated Safety Lid |
| For Use With (Equipment) | Culture Bottles and Vacuum Tubes |
| Quantity | 80 per Case |
| Volume (Metric) | 16 mL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.