Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAMSN1 Monoclonal Antibody (OTI1F6), TrueMAB™, OriGene
SDP

Catalog No. 501682447
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-2447 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-2447 Supplier OriGene Supplier No. TA808889
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SAMSN (HACS1) is a member of a novel gene family of putative adaptors and scaffold proteins containing SH3 and SAM (sterile alpha motif) domains. It encodes a 441 amino acid protein that is differentially expressed in hematopoietic cells and maligcies including myeloid leukemia, lymphoma, and myeloma. SAMSN has restricted expression in human tissues. It has been shown to be up-regulated by B cell activation signals and is a participant in B cell activation and differentiation. SAMSN is an immunoinhibitory adaptor that might be a useful target for immune suppression therapy.

Specifications

Antigen SAMSN1
Applications Western Blot
Classification Monoclonal
Clone OTI1F6
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene SAMSN1
Gene Accession No. Q9NSI8
Gene Alias HACS1, NASH1, SASH2, SH3D6B, SLy2
Gene Symbols SAMSN1
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 1-250 of human SAMSN1 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 64092
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.