Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sand, Reagents

Catalog No. 01337077
Change view
Click to view available options
Quantity:
500 g
3 kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-337-077 500 g
01-337-078 3 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-337-077 Supplier Reagents Holdings Llc Supplier No. C2299000500A5
Add to Cart
Add to Cart

Sand, Technical, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 14808-60-7
Molecular Formula O2Si
Molecular Weight (g/mol) 60.08
InChI Key VYPSYNLAJGMNEJ-UHFFFAOYSA-N
IUPAC Name silanedione
SMILES O=[Si]=O

Specifications

Boiling Range 2230°C
Melting Point 1610°C
Color White to Light Brown
Boiling Point 2230°C
Melting Range 1610°C
Packaging Poly Bottle
Quantity 500 g
Specific Gravity 2.4
Grade Technical
Physical Form Solid
Chemical Name or Material Sand
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.