Learn More
Essendant Foodservice Surface Sanitizer, Fragrance Free, 1 gal Bottle, 4/Carton
Supplier:  Essendant GOJ434104
Encompass_Preferred
Formulated for convenience and ease-of-use, Foodservice Surface Sanitizer Spray provides powerful germ-kill on the surfaces people touch most ¿ yet is gentle enough to use on a child¿s highchair with no rinse required. Patented fragrance-free formulation sanitizes without any harsh chemicals. It eliminates 99.9 percent of viruses and bacteria on surfaces ¿ including norovirus, E. coli, Salmonella, and human coronavirus in 30 seconds. With the EPA¿s lowest allowable toxicity rating, it delivers the overall fastest disinfection time of any Design for the Environment (DfE) product.
Catalog No. 27-901-83
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.