Learn More
Selleck Chemical LLC Santacruzamate A (CAY10683) 10mg 1477949-42-0
SDP
Supplier:  Selleck Chemical LLC S759510MG
Encompass_Preferred
Santacruzamate A (CAY10683) is a potent and selective HDAC inhibitor with IC50 of 119 pM for HDAC2, >3600-fold selectivity over other HDACs. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-6356
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.