Learn More
Medchemexpress LLC HY-B1776A 100mg Medchemexpress, Spermidine (trihydrochloride) CAS:334-50-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYB1776A100MG
Encompass_Preferred
Medchemexpress, HY-B1776A 100mg Spermidine (trihydrochloride) CAS:334-50-9 Spermidine trihydrochloride maintains cell membrane stability, increases antioxidant enzymes activities, improving photosystem II (PSII), and relevant gene expression. Spermidine trihydrochloride significantly decreases the H2O2 and O2.- contents. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2711
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.