Learn More
Specifications
Specifications
| Gene ID (Entrez) | 23450 |
| Species | Human |
| Product Line | Novus |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| For Use With (Application) | This Peptide is Useful as a Blocking Peptide for NB100-1077 |
| Gene Alias | KIAA0017splicing factor 3 b, subunit 3, 130 kD, Pre-mRNA-splicing factor SF3b 130 kDa subunit, RSE1, SAP130SAP 130, SF3B130, SF3b130pre-mRNA splicing factor SF3b, 130 kDa subunit, Spliceosome-associated protein 130, splicing factor 3 B subunit 3, splicing factor 3 b, subunit 3, 130 kDa, STAF130 |
| Gene Symbol | SF3B3 |
| Quantity | 0.1 mg |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
