Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAP30 Polyclonal Antibody, Active Motif™
SDP

Catalog No. p-7188637
Encompass
Change view
Click to view available options
Quantity:
10 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-4108 10 μL
50-199-4107 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-199-4108 Supplier Active Motif Supplier No. 39732
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Signal-recognition-particle assembly has a crucial role in targeting secretory proteins to the rough endoplasmic reticulum membrane. SRP9 together with SRP14 and the Alu portion of the SRP RNA, constitutes the elongation arrest domain of SRP. The complex of SRP9 and SRP14 is required for SRP RNA binding.

Specifications

Antigen SAP30
Applications ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot
Classification Polyclonal
Concentration Conc. Not Determined
Conjugate Unconjugated
Formulation whole serum with 30% glycerol and 0.035% sodium azide; pH
Gene SAP30
Gene Accession No. O75446
Gene Alias 30 kDa Sin3-associated polypeptide; 30kDa; BOS_8570; Histone deacetylase complex subunit SAP30; LOC100356863; Sap30; sin3 associated polypeptide; sin3 associated polypeptide, 30kD; Sin3 corepressor complex subunit SAP30; Sin3A associated protein 30; Sin3A associated protein 30kDa; Sin3A-associated protein; Sin3A-associated protein, 30kDa; Sin3-associated polypeptide p30; sin3-associated polypeptide, 30 kDa; Sin3-associated polypeptide, 30kDa; Sin3-associated protein
Gene Symbols SAP30
Host Species Rabbit
Immunogen This SAP30 antibody (pAb) was raised against recombinant full-length human SAP30 protein.
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8819
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.