Learn More
Enzo Life Sciences SAP97 monoclonal antibody (RPI 197.4) (50µg)
SDP
Supplier:  Enzo Life Sciences ADIVAMPS005D
Encompass_Preferred
Clone: RPI 197.4. Host: Mouse. Isotype: IgG1. Immunogen: Recombinant rat SAP97 (N-terminal fragment). Source: Purified from ascites. Species reactivity: Human, Mouse, Rat, Dog. Applications: ICC, IHC, IP, WB Electron microscopy. Purity: Protein G affinity purified. Formulation: Liquid. In PBS, pH 7.2, containing 50% glycerol and 0.09% sodium azide.
Catalog No. 50-200-7686
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.