Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Saponin, From Saponegin Glycosides (Off-White Powder/Practical Grade), MP Biomedicals

Catalog No. ICN10285525 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN10285525 25 g
ICN10285580 100 g
ICN10285590 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN10285525 Supplier MP Biomedicals, Inc Supplier No. 0210285525
Add to Cart
Add to Cart

Plant origin, Saponegin glycosides

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 637-39-8
Molecular Formula C6H16ClNO3
Molecular Weight (g/mol) 185.648
InChI Key HHLJUSLZGFYWKW-UHFFFAOYSA-N
Synonym triethanolamine hydrochloride, tea-hydrochloride, triethanolammonium chloride, 2,2',2-nitrilotriethanol hydrochloride, ethanol, 2,2',2-nitrilotris-, hydrochloride, 2,2',2-nitrilotrisethanol hydrochloride, unii-r297uj9qdy, tris 2-hydroxyethyl ammonium chloride, tris 2-hydroxyethyl amine hydrochloride, r297uj9qdy
PubChem CID 101814
IUPAC Name 2-[bis(2-hydroxyethyl)amino]ethanol;hydrochloride
SMILES C(CO)N(CCO)CCO.Cl

Specifications

Color White
Quantity 25 g
Grade Practical
Physical Form Powder
Chemical Name or Material Saponin, Cell biology
EINECSNumber 232-462-6
Recommended Storage Store at room temperature (15° to 30°C)
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.