Learn More
IBL America sAPPα (Mouse/Rat) (highly sensitive)
Supplier:  IBL America 27419
Encompass_Preferred
Solid phase sandwich ELISA for the determination of Mouse/Rat sAPP in serum, EDTA plasma and cell culture supernatant. For research use only, not for use in diagnostic procedures.This kit can measure Mouse/Rat sAPP. Tetra Methyl Benzidine (TMB) is used as a coloring agent (Chromogen). The strengthof coloring is proportional to the quantities of Mouse/Rat sAPP.
Catalog No. 50-136-6525
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.