Description
- Manufactured from medical grade polypropylene
- Thin-walled for efficient heat transfer during amplification
- Available in different colors
Specifications
Specifications
| Color | Assorted |
| Volume (Metric) | 0.2 mL |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Domed Cap (Attached) |
| Autoclavable | Not Autoclavable |
| Closure Included | Yes |
| Disposable | Single-use |
| Product Type | PCR Tube with Cap |