Learn More
Azure Biosystems Sapphire FL Biomolecular Imager
SDP
Catalog No. IS4100AZ Shop All Azure Biosystems Products
Change view
Click to view available options
Camera:
Laser
Catalog No. Camera
IS4100AZ Laser
1 options
Catalog No. IS4100AZ Supplier Azure Biosystems Supplier No. IS4100
Add to Cart
Add to Cart
Request a Quote
Provides upgradable laser scanner with a wide field of view, selection of user-adjustable lasers and filters
  • Laser based scanning system with wide field of view supporting 375 nm excitation to 900 nm emission
  • Adjustable Z focus
  • 5 - 1000 micron resolution
  • 46 x 25 cm FOV
Specifications
Type BioImaging System
Camera Laser
Focus Distance 46 x 25 cm
Resolution 5 - 1000 micron