Learn More
Azure Biosystems Sapphire FL Plate Holder
SDP
Catalog No. IS4054AZ Shop All Azure Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
Sapphire FL
Catalog No. For Use With (Equipment)
IS4054AZ Sapphire FL
1 options
Catalog No. IS4054AZ Supplier Azure Biosystems Supplier No. IS4054
Add to Cart
Add to Cart
Fixture designed to hold up to four standard 96-well plates with optically clear bottoms for use with the Sapphire FL.
  • Fixture designed to hold up to four standard 96-well plates with optically clear bottoms for use with the Sapphire FL.
Specifications
Product Type Sample Plate Holder
For Use With (Equipment) Sapphire FL