Learn More
Azure Biosystems Sapphire FL Slide Holder
SDP
Catalog No. IS4053AZ Shop All Azure Biosystems Products
Change view
Click to view available options
For Use With (Equipment):
Sapphire FL
Catalog No. For Use With (Equipment)
IS4053AZ Sapphire FL
1 options
Catalog No. IS4053AZ Supplier Azure Biosystems Supplier No. IS4053
Add to Cart
Add to Cart
Fixture designed to hold up to 15 standard microscope slides for use with the Sapphire FL.
  • Fixture designed to hold up to 15 standard microscope slides for use with the Sapphire FL.
Specifications
Product Type Slide Holder
For Use With (Equipment) Sapphire FL