Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Sapphire Flat Cap Strip for PCR 8-Tube Strip

Catalog No. 07000542 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Blue
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color
07-000-542 Blue
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 07-000-542 Supplier Greiner Bio-One Supplier No. 373292
Add to Cart
Add to Cart
This item is not returnable. View return policy

For use with PCR 8-tube Strip

  • For 8-tube PCR strip
  • Flat
  • Ergonomic
  • Blue
TRUSTED_SUSTAINABILITY

Specifications

Color Blue
For Use With (Equipment) PCR 8-tube strip
Sterility Non-sterile
Product Type Cap Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.