Learn More
Greiner Bio-One Sapphire PCR 8-Tube Strip with Cap Strip
Catalog No. 07000699 Shop All Greiner Bio-One Products
Change view
Click to view available options
Volume (Metric) Working:
0.2 mL
Catalog No. Volume (Metric) Working
07-000-699 0.2 mL
1 options
Catalog No. 07-000-699 Supplier Greiner Bio-One Supplier No. 671241
Add to Cart
Add to Cart
Ideal for qPCR. Greiner Bio-One Sapphire PCR 8-Tube Strip with Cap Strip are optically clear.
  • Ideal for qPCR
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Volume (Metric) Working 0.2 mL
Product Line PCR 8-Tube strip
Closure Type Hinged