Learn More
Greiner Bio-One Sapphire PCR 8-Tube Strip with Attached Strip Cap
Catalog No. 07000556 Shop All Greiner Bio-One Products
Change view
Click to view available options
Volume (Metric):
0.2 mL
Catalog No. Volume (Metric)
07-000-556 0.2 mL
1 options
Catalog No. 07-000-556 Supplier Greiner Bio-One Supplier No. 608201
Add to Cart
Edge
Add to Cart
Greiner Bio-One Sapphire PCR 8-Tube Strip has thin walls for optimal heat transfer.
  • Non-pyrogenic
  • Free of detectable DNase, RNase, human DNA
  • Thin wall for optimal heat transfer
  • Low evaporation rate in PCR
  • With Attached Strip Cap
TRUSTED_SUSTAINABILITY
Specifications
Color Clear
Volume (Metric) 0.2 mL
Quantity 125/Pk.
Sterility Non-sterile
Closure Type Strip Cap (Attached)
Product Type PCR 8-Tube Strip