Learn More
Greiner Bio-One Sapphire PCR 8-Tube Strip with Individually Attached Cap
Shop All Greiner Bio-One ProductsDescription
- Non-pyrogenic
- Free of detectable DNase, RNase, human DNA
- Thin wall for optimal heat transfer
- Low evaporation rate in PCR
- Individually attached caps to minimise cross contamination
Specifications
Specifications
| Color | Natural |
| Volume (Metric) | 0.2 mL |
| Sterility | Non-sterile |
| Material | Polypropylene |
| Closure Type | Flat Cap (Attached) |
| Autoclavable | Not Autoclavable |
| Closure Included | Yes |
| Disposable | Single-use |
| Product Type | Thin Wall PCR Tubes with Caps (8-tube Combo Strip) |