Learn More
Greiner Bio-One Sapphire PCR 8-Tube Strip with Individually Hinged Caps
Catalog No. 07000698 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
White
Catalog No. Color
07-000-698 White
1 options
Catalog No. 07-000-698 Supplier Greiner Bio-One Supplier No. 671231
Add to Cart
Edge
Add to Cart
Ideal for qPCR. Greiner Bio-One Sapphire PCR 8-Tube Strip with Individually Hinged Caps are white with optically clear caps
  • Ideal for qPCR
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Color White
Volume (Metric) Working 0.2 mL
Product Line PCR 8-Tube strip
Closure Type Flat Cap (Attached)