Learn More
Greiner Bio-One Sapphire PCR 8-Tube Strip with Individually Hinged Caps - LOW PROFILE
Catalog No. 07000697 Shop All Greiner Bio-One Products
Change view
Click to view available options
Volume (Metric) Working:
0.1 mL
Catalog No. Volume (Metric) Working
07-000-697 0.1 mL
1 options
Catalog No. 07-000-697 Supplier Greiner Bio-One Supplier No. 671221
Add to Cart
Edge
Add to Cart
Ideal for qPCR. Greiner Bio-One Sapphire PCR 8-Tube Strip with Individually Hinged Caps - LOW PROFILE are optically clear
  • Ideal for qPCR
  • Low Profile
  • Free of detectable DNase, RNase and human DNA
  • Non-pyrogenic
TRUSTED_SUSTAINABILITY
Specifications
Closure Color Clear
Volume (Metric) Working 0.1 mL
Product Line PCR 8-Tube strip
Closure Type Hinged (Individually Attached)
Product Type Low Profile