Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Greiner Bio-One Sapphire RT-PCR 8-Tube Strips and Caps Combo

Catalog No. 07000713 Shop All Greiner Bio-One Products
Change view
Click to view available options
Color:
Clear
White
Format:
Low Profile
Standard Profile
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Format
07-000-713 Clear Low Profile
07-000-712 Clear Standard Profile
07-000-714 White Low Profile
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 07-000-713 Supplier Greiner Bio-One Supplier No. 673281
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For use with real-time/quantitative PCR. Greiner Bio-One Sapphire RT-PCR 8-Tube Strips and Caps Combo is compatible with comprehensive thermal cycler.

  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
TRUSTED_SUSTAINABILITY

Specifications

Color Clear
Volume (Metric) 0.2 mL
For Use With (Equipment) Real-time/quantitative PCR
Sterility Non-sterile
Material Polypropylene
Format Low Profile
Product Type PCR 8-Tube Strips and Cap Combo
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.