Learn More
Greiner Bio-One Sapphire RT-PCR 8-Tube Strips and Ergonomic Caps Combo
Catalog No. 07000719 Shop All Greiner Bio-One Products
Change view
Click to view available options
Closure Type:
Flat Strip Cap
Optical Strip Cap
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Closure Type
07-000-719 Optical Strip Cap
07-000-718 Flat Strip Cap
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 07-000-719 Supplier Greiner Bio-One Supplier No. 673287
Add to Cart
Add to Cart
This item is not returnable. View return policy
For use with real-time/quantitative PCR. Greiner Bio-One Sapphire RT-PCR 8-Tube Strips and Ergonomic Caps Combo are compatible with comprehensive thermal cycler.
  • Thin-walled for heat transfer during amplification
  • Free of detectable DNase, RNase, human DNA, endotoxin and PCR inhibitors
  • Packaging labelled with product symbols for warehousing
  • Used for sample handling
TRUSTED_SUSTAINABILITY
Specifications
Volume (Metric) 0.2 mL
For Use With (Equipment) Real-time/quantitative PCR
Sterility Non-sterile
Material Polypropylene
Closure Type Optical Strip Cap
Product Type RT-PCR 8-Tube Strips and Caps Combo