Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SAR1B Mouse anti-Human, Mouse, Porcine, Rabbit, Rat, Clone: 1B11A1, ProteinTech
SDP

Catalog No. 502239817 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-223-9817 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 50-223-9817 Supplier Proteintech Group Inc Supplier No. 677651IG150UL
Only null left
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Aliquoting is unnecessary for -20°C storage.

SAR1B Monoclonal antibody specifically detects SAR1B in Human, Mouse, Porcine, Rabbit, Rat samples. It is validated for Western Blot

Specifications

Antigen SAR1B
Applications Western Blot
Classification Monoclonal
Clone 1B11A1
Concentration 1000 μg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SAR1B
Gene Accession No. Q5HZY2, Q5PYH3, Q9CQC9, Q9Y6B6
Gene Alias 2310075M17Rik; 2900019I22Rik; ANDD; CMRD; GTBPB; GTP-binding protein B; GTP-binding protein SAR1b; GTP-binding protein Sara; SAR1 gene homolog B; SAR1 gene homolog B (S. cerevisiae); SAR1 homolog B; SAR1a gene homolog 2; Sar1b; SAR1b gene homolog; SARA1B; SARA2; SARB; secretion associated Ras related GTPase 1B; secretion associated, Ras related GTPase 1B
Gene Symbols SAR1B
Host Species Mouse
Immunogen SAR1B Fusion Protein Ag7718
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100340384, 287276, 494017, 51128, 66397
Target Species Human, Mouse, Pig, Rabbit, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.