Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Sarcalumenin Monoclonal Antibody (XIIC4)
GREENER_CHOICE

Catalog No. MA3932 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA3932 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA3932 Supplier Invitrogen™ Supplier No. MA3932
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Mouse Monoclonal Antibody

MA3-932 detects sarcalumenin from canine, guinea pig, mouse and rabbit tissues. This antibody specifically detects sarcalumenin and the 53 kDa glycoprotein splice variant. MA3-932 has been successfully used in Western blot. By Western blot, this antibody detects a 53 kDa and a 160 kDa protein representing the glycoprotein and sarcalumenin, respectively, in rabbit skeletal muscle extracts. The MA3-932 antigen is purified rabbit sarcoplasmic reticulum membrane vesicles.

Sarcalumenin is a 160 kDa acidic calcium binding glycoprotein found to reside at the inner membrane of the sarcoplasmic reticulum in a calcium dependent manner. The gene encoding sarcalumenin also encodes an alternate splice variant, a 53 kDa glycoprotein which co-localizes with sarcalumenin. These two proteins are found in the longitudinal sarcoplasmic reticulum and the nonjunctional membranes of the terminal cisternae. They have been observed to co-localize with calcium-ATPase suggesting that they are involved in calcium transport and sequestration.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Sarcalumenin
Applications Western Blot, Immunocytochemistry
Classification Monoclonal
Clone XIIC4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene SRL
Gene Accession No. P13666, Q7TQ48
Gene Alias 9830004M20Rik; AW108387; Sar; sarcalumenin; sarcolumenin; sarcolumenin precursor; sarcoplasmic reticulum glycoprotein; Srl
Gene Symbols SRL
Host Species Mouse
Immunogen Purified rabbit sarcoplasmic reticulum membrane vesicles.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009112, 100719273, 106393, 302948, 490030
Target Species Mouse, Rat, Canine, Rabbit, Guinea Pig
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.