Learn More
Invitrogen™ Sarcomeric alpha Actinin Monoclonal Antibody (EA-53)
Shop All Thermo Scientific Products

Description
MA1-22863 detects sarcomeric alpha Actinin in bovine, feline, chicken, fish, goat, hamster, lizard, mouse porcine, rabbit, rat, ovine, snake, and Xenopus laevis samples.
Specifications
Specifications
| Antigen | Sarcomeric alpha Actinin |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Free Floating), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | EA-53 |
| Concentration | Conc. Not Determined |
| Conjugate | Unconjugated |
| Formulation | Ascites with 0.09% sodium azide |
| Gene | ACTN1 |
| Gene Accession No. | P12814, Q7TPR4, Q9Z1P2 |
| Gene Alias | 3110023F10Rik; actinin 1 smooth muscle; actinin alpha 1; actinin, alpha 1; ACTN1; Actn1a; alpha actinin 1a; alpha-actinin cytoskeletal isoform; Alpha-actinin-1; BDPLT15; F-actin cross-linking protein; non-muscle alpha-actinin 1; non-muscle alpha-actinin-1 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.