Learn More
MyBioSource SARM ELISA Kit
SDP
Supplier:  MyBioSource MBS931953196STRIPWELLS
Encompass_Preferred
The MyBioSource SARM ELISA Kit is an enzyme-linked immunosorbent assay for the quantitative measurement of SARM in serum, plasma, tissue homogenates, cell lysates, cell culture supernates and other biological fluids. The kit uses a double-antibody sandwich ELISA to assay SARM levels. It has a detection range of 0.156-10 ng/ml SARM. The sensitivity of this assay is < 0.094 ng/ml. The assay takes around 2 hours to complete. This ELISA kit shows no cross-reactivity with other cytokines. It includes a micro ELISA plate, standard, antibody, HRP-conjugate, TMB substrate, wash buffer and other reagents and materials needed to run the ELISA. This is a high quality ELISA kit from a reliable supplier.
Catalog No. NC2037968
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.