Learn More
TARGETMOL CHEMICALS INC SAROGLITAZAR MAGNESIUM 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T128345MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Saroglitazar Magnesium is a novel peroxissome proliferator-activated receptor (PPAR) agonist with significant activation activity against PPAR(alpha) (EC50 0.65pM) and moderate activation against PPAR(gamma) (EC50 3 nM). purity: 98%
Catalog No. 50-258-1417