Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS/SARS-CoV-2 Nucleocapsid Monoclonal Antibody (B46F)
GREENER_CHOICE

Catalog No. PIMA17404 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA17404 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA17404 Supplier Invitrogen™ Supplier No. MA17404
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1-7404 is reactive with the NP of SARS-CoV-2 (COVID-19 virus) by ELISA. Negative for human coronaviruses 229E & OC43. ELISA dilution range suggested 1:20- 1:200. The MA1-7404 antibody reacts with SARS Coronavirus nucleoprotein in viral and infected tissue samples. MA1-7404 has been successfully used in ELISA, Western blot and Immunofluorescence applications. The MA1-7404 antibody was raised against a SARS virus nucleoprotein preparation.

SARS-CoV-2 Nucleocapsid Protein is associated with nucleic acid. It is the most abundant protein for coronavirus. Because of the strong immunogenicity of coronavirus Nucleocapsid, it is believed that SARS-CoV-2 Nucleocapsid Protein has potential value for the diagnosis of the virus. The SARS-CoV-2 N protein is an abundant RNA-binding protein critical for viral genome packaging, yet the molecular details that involve this process are poorly understood. N protein contains three dynamic disordered regions that house putative transiently-helical binding motifs. The two folded domains interact minimally such that full-length N protein is a flexible and multivalent RNA-binding protein. SARS-CoV-2 causes illnesses ranging from the common cold to severe diseases.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS/SARS-CoV-2 Nucleocapsid
Applications ELISA, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone B46F
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.2
Gene N
Gene Accession No. P0DTC9
Gene Alias COVID19 Nucleocapsid; COVID-19 nucleocapsid; COVID19 Spike nucleocapsid; SARS-CoV-2 N
Gene Symbols N
Host Species Mouse
Immunogen SARS nucleoprotein preparation.
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 43740575
Target Species Virus
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.