Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS Coronavirus Spike Protein Recombinant Mouse Monoclonal Antibody (17F706R)
GREENER_CHOICE

Catalog No. 01676482
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-676-482 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-676-482 Supplier Invitrogen™ Supplier No. MA141173
Only null left
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

The immunogen is 100% (16/16) homologous to SARS-COV-2 (COVID-19) Spike protein. The cross reactivity is not empirically tested. This is the mouse recombinant monoclonal version of the clone 17F706 produced from mouse ascites. Immunogen sequence: QPELDSFKEELDKYFKN.

It has recently been shown that SARS (severe acute respiratory syndrome) is caused by a human coronavirus. Human coronaviruses are the major cause of upper respiratory tract illness, such as the common cold, in humans. Coronaviruses are positive-stranded RNA viruses, featuring the largest viral RNA genomes known to date (27-31 kb). The first step in coronavirus infection is binding of the viral spike protein, a 139-kDa protein, to certain receptors on host cells. The spike protein is the main surface antigen of the coronavirus. The glycosylated spike protein (as well as the nucleocapsid protein) can be detected in infected cell culture supernatants with antisera from SARS patients.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS Coronavirus Spike Protein
Applications Western Blot
Classification Recombinant Monoclonal
Clone 17F706R
Concentration 1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Alias Coronavirus; COVID; Covid 19; COVID19; COVID-19; SARS; SARS Coronavirus; SARS CoV; SARS CoV2; SARS CoV-2; SARS Spike Protein; SARS-CoV2; SARS-CoV-2
Host Species Mouse
Immunogen Synthetic peptide corresponding to amino acids 1124-1140 from the S (Spike glycoprotein) (Spike protein S2') for the Human SARS coronavirus.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.