Learn More
Description
Specifications
Specifications
| For Use With (Application) | Bioactivity |
| Formulation | 4 mM in DMSO |
| Gene ID (Entrez) | 43740578 |
| Molecular Weight (g/mol) | M.W.-theoretical: 1096.3 Da |
| Quantity | 200 μg |
| Storage Requirements | Protect from light. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 60 months from date of receipt, -20°C to -70°C as supplied. 3 months, -20°C to -70°C under sterile conditions after opening. |
| Gene Alias | 3 CL Proteinase, 3 C-like main protease, 3 CL-Mpro, COVID-19, M Proteinase, Main Protease, Mpro |
| Purity or Quality Grade | >95%, by HPLC. 1096.6 Da |
| Protein | SARS-CoV-2 3 CL Protease |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
