Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 3CLpro Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116940 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116940 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116940 Supplier Invitrogen™ Supplier No. PA5116940
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

Positive Samples: 293T Immunogen sequence: MESLVPGFNE KTHVQLSLPV LQVRDVLVRG FGDSVEEVLS EARQHLKDGT CGLVEVEKGV LPQLEQPYVF IKRSDARTAP HGHVMVELVA ELEGIQYGRS GETLGVLVPH VGEIPVAYRK VLLRKNGNKG AGGHSYGADL KSFDLGDELG TDPYEDFQEN WNTKHSSGVT RELMRELNGG AYTRYVDNNF CGPDGYPLEC IKDLLARAGK ASCTLSEQLD FIDTKRGVYC CREHEHEIAW YTERSEKSYE LQTPFEIKLA KKFDTFNGEC PNFVFPLNSI IKTIQPRVEK KKLDGFMGRI RSVYPV.

Severe Acute Respiratory Syndrome (SARS), an emerging disease characterized by atypical pneumonia, has been attributed to a novel coronavirus (SARS-CoV). The SARS 3C-like protease (SARS 3CL (pro)) is a cysteine protease engaging in the proteolytic cleavage of the viral precursor polyprotein to a series of functional proteins required for corona virus replication and is considered an attractive target for therapeutics against SARS.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SARS-CoV-2 3CLpro
Applications ELISA, Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.98 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Alias Coronavirus; COVID; COVID 19; COVID-19; SARS Coronavirus; SARS CoV; SARS CoV2; SARS-CoV-2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.