Learn More
Genetex Inc SARS-CoV-2 (COVID-19) RdRp (nsp12) antibody | 100μl
SDP
Supplier:  Genetex Inc GTX135467
Encompass
Carrier-protein conjugated synthetic peptide encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) RdRp (nsp12) (Strain:Wuhan-Hu-1). The exact sequence is proprietary.
Catalog No. NC1860823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.