Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein NTD (A262S), His Tag
Supplier:  ACROBiosystems S1DC52H5100UG
Encompass_Preferred
Recombinant Protein;100UG;SARS-CoV-2 S1 protein NTD (A262S), His Tag (S1D-C52H5) is expressed from human 293 cells (HEK293). It contains AA Ser 13 - Leu 303 (Accession # QHD43416.1 (A262S)).;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 34.9 kDa.
Catalog No. 50-205-4749
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.