Learn More
ACROBiosystems SARS-CoV-2 (COVID-19) S1 protein (HV69-70del), His Tag
Supplier:  ACROBiosystems S1NC52HD1MG
Encompass_Preferred
Recombinant Protein;1MG;SARS-CoV-2 S1 protein (HV69-70del), His Tag (S1N-C52Hd) is expressed from human 293 cells (HEK293). It contains AA Val 16 - Arg 685 (Accession # QHD43416.1 (HV 69-70del)). HV69-70 deletion was identified in the SARS-CoV-2 variant (known as 20B/501Y.V1, VOC 202012/01, or B.1.1.7 lineage) which emerged in the United Kindom.;This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 76.6 kDa.
Catalog No. 50-205-4785
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.