Learn More
Johannes Lieder SARS-CoV-2 Covid-19 Tissues on Microscope Slides
SCI_ED
Catalog No. S139705 Shop All Johannes Lieder GmbH & Co. KG Products
Change view
Click to view available options
Mounting Type:
Cross and Longitudinal Section
Catalog No. Mounting Type
S139705 Cross and Longitudinal Section
1 options
This item is not returnable. View return policy
Catalog No. S139705 Supplier Johannes Lieder Supplier No. SARSCOV2
Add to Cart
Add to Cart
This item is not returnable. View return policy
10 microscope slides of affected organs on the human body by SARS-CoV-2 infection.
  • 10 microscope slides with accompanying booklet
  • Targets of the virus:
    1. Ma112c Stratified non-cornified squamous epithelium in section through buccal gum
    2. Ho607e Cornea from eye human t.s.
    3. Ho612f Olfactory epithelium human t.s.Disturbance of the sense of taste is the most common symptom
    4. Ho610f Wallate papillae with taste buds human t.s. Important organ of immune defence in the throat
    5. Ho233e Tonsil (Tonsilla palatina) human t.s. Structure of the lower respiratory tract
    6. Ho214f Trachea human t.s.
    7. Ho220e Bronchus of lung human t.s.
    8. Ho216e Lung human sec. routine stained
    9. Ma217d Lung of cat t.s. stained for elastic fibres.
    10. Frequent secondary infection with severe courses or previous diseases
    11. 9218e Influenzal pneumonia
Specifications
Classification Respiratory System
Material Elastin Stained Glass
Mounting Type Cross and Longitudinal Section
Slide Type Prepared Slide