Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SARS-CoV-2 Envelope (aa1-75), His-Avi Tag Recombinant Protein

Catalog No. PIRP87682
Encompass
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIRP87682 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIRP87682 Supplier Invitrogen™ Supplier No. RP87682
Add to Cart
Edge
Add to Cart

Recombinant Protein

Description: Recombinant 2019-nCoV envelope Protein is produced by E. coli expression system.

The SARS-CoV-2 envelope (E) protein is a small, integral membrane protein involved in several aspects of the virus' life cycle, such as assembly, budding, envelope formation, and pathogenesis.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 0.47 mg/mL
For Use With (Application) Control, ELISA, Western Blot
Formulation 20 mM tris with 0.5% Triton X-100, 250 mM NaCl and no preservative; pH 8
Gene ID (Entrez) 43740570
Molecular Weight (g/mol) 11.4 kDa
Name SARS-CoV-2 Envelope (aa1-75), His-Avi Tag
Quantity 100 μg
Source E. coli
Regulatory Status RUO
Endotoxin Concentration <0.1 EU/ μg
Gene Alias COVID19 Envelope Protein
Common Name SARS-CoV-2 Envelope Protein
Biological Activity Measured by its binding ability in a functional ELISA. Immobilized Recombinant SARS-CoV-2 Envelope (at 2 μg/mL (100 μL/well) can bind Recombinant SARS-CoV-2 Nucleocapsid with a linear range of 1.2-41.1 ng/mL)
Conjugate Unconjugated
Recombinant Recombinant
Protein Tag His-tag, Avi-tag
Sequence SARS-CoV-2 Envelope Protein, amino acids Met1-Val75 (Accession # QHD43418.1)
Content And Storage -80°C
Expression System E. coli
Form Liquid
Purity or Quality Grade >95% by SDS-PAGE
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.