Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abcam SARS-CoV-2 Envelope Protein Peptide, Synthetic, Blocking, 50 μg

Catalog No. 502704041
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-270-4041 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-270-4041 Supplier Abcam Supplier No. AB27306450UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Synthetic blocking peptide for use in blocking experiments.

  • Protein Name: SARS-CoV-2 envelope protein
  • Type: Synthetic blocking peptide
  • Applications: BL
  • Form: Liquid
  • Concentration: 0.2 mg/mL
  • Storage Buffer: PBS, 0.1% BSA, 0.02% Sodium azide, pH 7.2
  • Storage Conditions: Store at -20°C

The product is subject to the following: Abcam Restricted Use Statement

Specifications

Target Molecule SARS-CoV-2 envelope protein
Substrate Properties Peptide-based Substrate
Shipping Condition Blue Ice
Reactivity HCoV
Product Type Blocker and Diluent
Buffer PBS, 0.1% BSA, 0.02% sodium azide, pH7.2
Concentration 0.2 mg/mL
Content And Storage Store at -20°C.
Form Liquid
Label or Dye Unconjugated
Cross Reactivity SARS-CoV-2
Formulation PBS, 0.1% BSA, 0.02% Sodium azide, pH 7.2
Quantity 50 μg
For Use With (Application) Blocking
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.