Learn More
AdipoGen SARS-CoV-2 Inhibitor Screening Kit (96 wells)
SDP
Supplier:  AdipoGen AG48B0001KI01
Encompass_Preferred
Screening Assay: To screen compounds that inhibit the binding of the viral protein SARS-CoV-2 Spike Protein (RBD) to its human receptor ACE2. SARS-CoV-2 contains 4 structural proteins, including Envelope (E), Membrane (M), Nucleocapsid (N) and Spike (S), which is a transmembrane protein, composed of two subunits S1 and S2. The S1 subunit contains a receptor binding domain (RBD), which binds to the cell surface receptor Angiotensin-Converting Enzyme 2 (ACE2) present at the surface of epithelial cells, causing mainly infection of human respiratory cells. The SARS-CoV-2 Inhibitor Screening Kit contains key reagents required to facilitate identification of SARS-CoV-2 inhibitors, based on the binding of the RBD of the Spike S protein from SARS-CoV-2 to its human receptor ACE2. This assay allows to identify and characterize the effect of different inhibitory molecules including antibodies or chemicals on the prevention of binding of SARS-CoV-2 virus to any ACE2-expressing cells.
Catalog No. 50-205-3345
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.