Learn More
ZeptoMetrix SARS-CoV-2 (Isolate: USA-WA1/2020) Lysate
SDP
Catalog No. 22158375 Shop All ZeptoMetrix Corporation Products
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
22-158-375 1 mg
22-158-376 0.5 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 22-158-375 Supplier ZeptoMetrix Supplier No. 0810587
Add to Cart
Add to Cart
SARS-CoV-2 (Isolate: USA-WA1/2020) Culture Fluid is an enveloped, positive-sense single-stranded RNA virus from the Coronaviridae family and the Betacoronaviridae genus.
  • This product is intended for research, product development, quality assurance testing, or further manufacturing use.
  • Viral lysates can be used as an antigen, as a source for the purification of viral proteins, or in assays for the detection of viral antibodies.
Specifications
Content And Storage Recommended storage temperature -65°C or below
Type Lysate
Intended Use For research or further manufacturing only. Not for use in diagnostic procedures
Quantity 1 mg