Learn More
Invitrogen™ SARS-CoV-2 NSP12 Recombinant Rabbit Monoclonal Antibody (HL1444)

Description
Store as concentrated solution. Centrifuge briefly prior to opening vial.
Specifications
Specifications
| Antigen | SARS-CoV-2 NSP12 |
| Applications | Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Clone | HL1444 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative |
| Gene Alias | COVID19 Nsp12; COVID-19 Nsp12; SARS-CoV-2 Nonstructural Protein 12; SARS-CoV2 NSP12; SARS-COV-2 NSP12 |
| Host Species | Rabbit |
| Immunogen | Recombinant protein encompassing a sequence within the center region of SARS-CoV-2 (COVID-19) nsp12 / RdRP. The exact sequence is proprietary. |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.